Learn More
Bachem Z-Pyr-OSu, 25g, 40291-26-7, MFCD00191144 DFS Item
Manufacturer:  Bachem 4015205.0025
Also called 5-oxoproline or pidolic acid. Bachem uses the acronym Pyr for denoting pyroglutamic acid. Further abbreviations as Glp, Pgl, pGlu (pE), or < Glu (< E) are also in use.
Catalog No. 50-194-7300