Learn More
Mac Mod AnalyticalSupplier Diversity Partner ALT 5 C8 250X4.6MM DFS Item
Manufacturer:  Mac Mod AnalyticalSupplier Diversity Partner 88076
Alltima 5um C8 Column 250x4.6mm Note: All Grace Analytical and Semi-Preparative HPLC Columns brands including Vydac®, Alltima®, Alltima® HP, Prevail™, Apollo™, Allsep®, Apex™ and Genesis™ analytical HPLC column ranges are now available in the US from MAC-MOD Analytical via Fisher Scientific - Hichrom Limited recently obtained worldwide manufacturing rights to these brands and is offering them in the US through MAC-MOD Analytical. These columns are made with the same exact manufacturing protocols and to the identical specifications previously used by Grace. Category: Chromatography Columns and Supplies GRACE Part Number: 88076 Alternative Part Number: 5129313
Catalog No. 50-112-0377