Learn More
Mac Mod AnalyticalSupplier Diversity Partner ALT HP 3 HILIC 50X2.1MM DFS Item
Manufacturer:  Mac Mod AnalyticalSupplier Diversity Partner 86461
Alltima HP 3um HILIC Column 50x2.1mm Note: All Grace Analytical and Semi-Preparative HPLC Columns brands including Vydac®, Alltima®, Alltima® HP, Prevail™, Apollo™, Allsep®, Apex™ and Genesis™ analytical HPLC column ranges are now available in the US from MAC-MOD Analytical via Fisher Scientific - Hichrom Limited recently obtained worldwide manufacturing rights to these brands and is offering them in the US through MAC-MOD Analytical. These columns are made with the same exact manufacturing protocols and to the identical specifications previously used by Grace. Category: Chromatography Columns and Supplies GRACE Part Number: 86461 Alternative Part Number: 5177760
Catalog No. 50-112-0769