Learn More
AdipoGenSupplier Diversity Partner ALTERNARIOLMONOME ETHER(5MG) DFS Item
Manufacturer:  AdipoGenSupplier Diversity Partner BVT0323M005
Alternariol monomethyl ether, AdipoGen Life Sciences. Chemical. CAS: 26894-49-5. Formula: C15H12O5. MW: 272.3. Isolated from Alternaria sp. Mycotoxin. Antifungal. Phytotoxic. Shown to possess DNA strand-breaking activity. Inhibits the activity of topoisomerase IIalpha (Topo IIalpha) in mammalian cells. Antiviral (Herpes simplex). Hepatitis C NS3-4A protease inhibitor.
Catalog No. 50-114-6805