Learn More
Swift™ Altimeter/Barometer Products for Science Education
Visually demonstrates the connection between altitude and atmospheric pressure to help your students understand this relationship.
Manufacturer:  Swift™ 478
Catalog No. S41544A
Add to cart
Includes: A zipper pouch is included.
Description
Description
  • Portable, ideal for field trips
  • Barometric pressure decreases as the altitude above sea level increases, atmospheric barometers can be used to measure the height above sea level.
  • Can be mounted or carried in a pocket
Order Info
Measures 2 1/2 × 2 1/2 × 1 in..
Specifications
Specifications
Visually demonstrates the connection between altitude and atmospheric pressure to help your students understand this relationship
Zipper pouch
Altimeter/Barometer