Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Altretamine, USP, 98-102%, Spectrum™

Click to view available options
Quantity:
1 kg
5 kg
Packaging:
Poly Bottle
Poly Pail
645-05-6
C9H18N6
210.29
UUVWYPNAQBNQJQ-UHFFFAOYSA-N
N2,N2,N4,N4,N6,N6-hexamethyl-1,3,5-triazine-2,4,6-triamine
CN(C)C1=NC(=NC(=N1)N(C)C)N(C)C
645-05-6
C9H18N6
210.29
UUVWYPNAQBNQJQ-UHFFFAOYSA-N
N2,N2,N4,N4,N6,N6-hexamethyl-1,3,5-triazine-2,4,6-triamine
CN(C)C1=NC(=NC(=N1)N(C)C)N(C)C

A3035, 645-05-6, C9H18N6

Spectrum™ Altretamine, USP is used to ease the symptoms of ovarian cancer. All Spectrum Chemical manufactured USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Packaging Poly Bottle
Ignition Residue 0.001
Water 0.01
Quantity 1 kg
CAS Max % 1
Percent Purity 98 to 102%