Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Brady™ Alum Sign: CAUTION: H2S POIS. GAS MAY BE PRESENT. NO OPEN FLAMES. WEAR NECES. PROTECT. EQUIP AND PROCEED w/ CAUTION
Improve workplace safety with proper signage and markings. This Brady™ aluminum sign is a durable, clear and effective tool to raise awareness to workplace hazards.
Specifications
Material | Aluminum |
---|---|
Shape | Rectangular |
Displayed Text | CAUTION H2S POISONOUS GAS MAY BE PRESENT. NO OPEN FLAMES. WEAR NECESSARY PROTECTIVE EQUIPMENT AND PROCEED WITH CAUTION. |
Catalog Number | Mfr. No. | Dimensions (L x W) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Dimensions (L x W) | Price | Quantity & Availability | |||||
19-106-541
|
Brady™
145070 |
14 x 10 in. | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
19-106-543
|
Brady™
145072 |
10 x 7 in. | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
Description
- Aluminum material is abrasion-resistant and chemical-resistant
- Safety signs are important facility visuals that are vital to communicating warnings and other safety information
- Aluminum signs provide the strength and durability of metal, but never rust and can tolerate extreme temperatures, years of chemical spills, wash downs and cleanings
- Bold and bright, providing high visibility to critical messages
- For indoor and outdoor use
Specifications
Aluminum | |
Rectangular | |
CAUTION H2S POISONOUS GAS MAY BE PRESENT. NO OPEN FLAMES. WEAR NECESSARY PROTECTIVE EQUIPMENT AND PROCEED WITH CAUTION. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title