Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alum Solution, 10% (w/v), For Dissolved Oxygen, APHA, Spectrum™

Click to view available options
Quantity:
500 mL
1 L
Packaging:
Poly Bottle
7784-24-9
7784-24-9
Grade Chemical Solution
Packaging Poly Bottle
Quantity 1 L