Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminium, 99.9995%, Acros Organics™

Click to view available options
Quantity:
25g
Packaging:
Glass bottle
7429-90-5
Al
26.982
XAGFODPZIPBFFR-UHFFFAOYSA-N
aluminium,metal,powder,aluminio,metana,adom,alumina fibre,aluminium flake,dust,noral aluminium
5359268
CHEBI:33629
aluminum
[Al]
7429-90-5
Al
26.982
XAGFODPZIPBFFR-UHFFFAOYSA-N
aluminium,metal,powder,aluminio,metana,adom,alumina fibre,aluminium flake,dust,noral aluminium
5359268
CHEBI:33629
aluminum
[Al]

For ultra trace analysis

For ultra trace analysis
Specific Gravity 2.7
Quantity 25g
Density 2.7000g/mL
Assay Percent Range 99.9995%
Packaging Glass bottle
Formula Weight 26.97
Physical Form Shot
Percent Purity 99.9995%
Grade Trace Metal Basis
Total Trace Metal Impurities 5ppm max.
Color Gray
Melting Point 660.0°C
Boiling Point 2327.0°C
Chemical Name or Material Aluminium
Show More Show Less