Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Aluminum ammonium sulfate dodecahydrate, 99%, pure, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
100 g
1 kg
2.5 kg
10 kg
Packaging:
Glass Bottle
Plastic bottle
Plastic drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Boiling Point | 200.0°C |
Melting Point | 93.5°C |
Color | White |
Physical Form | Crystals |
Quantity | 1 kg |
Assay Percent Range | 99% |
Molecular Formula | AlH28NO20S2 |
Linear Formula | AlNH4(SO4)2·12H2O |
Solubility Information | Solubility in water: 150g/L (20°C). Other solubilities: freely soluble in glycerol, practically insoluble in alcohol |
Molecular Weight (g/mol) | 453.313 |
Show More |
Safety and Handling
Hazard Statement | GHS H Statement: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | GHS P Statement: Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification. |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction