Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminium ammonium sulfate dodecahydrate, 99%, pure, ACROS Organics™

Click to view available options
Quantity:
5g
100g
1kg
2.5kg
10kg
Packaging:
Glass Bottle
Plastic bottle
Plastic drum
7784-26-1
H4AlNO8S2·12H2O
453.33
MFCD00149958
WZUKKIPWIPZMAS-UHFFFAOYSA-K
ammonia alum,unii-5c36drl9zn,aluminium ammonium sulfate dodecahydrate,aluminum ammonium disulfate dodecahydrate,ammonium aluminum sulfate hydrate,alum, ammonium usp,aluminum ammonium sulfate dodecahydrate,ammonium aluminum sulfate dodecahydrate,5c36drl9zn,ammonium aluminum disulfate dodecahydrate
62668
aluminum;azanium;disulfate;dodecahydrate
[NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Al+3]
7784-26-1
H4AlNO8S2·12H2O
453.33
MFCD00149958
WZUKKIPWIPZMAS-UHFFFAOYSA-K
ammonia alum,unii-5c36drl9zn,aluminium ammonium sulfate dodecahydrate,aluminum ammonium disulfate dodecahydrate,ammonium aluminum sulfate hydrate,alum, ammonium usp,aluminum ammonium sulfate dodecahydrate,ammonium aluminum sulfate dodecahydrate,5c36drl9zn,ammonium aluminum disulfate dodecahydrate
62668
aluminum;azanium;disulfate;dodecahydrate
[NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Al+3]
Assay Percent Range 98.5% min. (Complexometry)
Linear Formula AlNH4(SO4)2·12H2O
Merck Index 15,323
Solubility Information Solubility in water: 150g/L (20°C). Other solubilities: freely soluble in glycerol,practically insoluble in alcohol
Formula Weight 453.33
Physical Form Crystals
Percent Purity 99%
Grade Pure
Density 1.6400g/mL
Loss on Drying 45% to 48% (2 g, 300°C)
Packaging Plastic bottle
Solubility Solubility in water: 150g/L (20°C). Other solubilities: freely soluble in glycerol, practically insoluble in alcohol
Specific Gravity 1.64
Boiling Point 200.0°C
Color White
Melting Point 93.5°C
Quantity 100g
Chemical Name or Material Aluminium ammonium sulfate dodecahydrate
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning