Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminium chloride, 99.999%, (trace metal basis), for analysis, anhydrous, powder, ACROS Organics™

Click to view available options
Quantity:
10g
20g
100g
Packaging:
Glass bottle
7446-70-0
AlCl3
133.34
aluminium chloride anhydrous,aluminum iii chloride,aluminiumchloride,aluminumchloride,aluminum chlorid,aluminum cloride,alumimum chloride,aluminum-chloride,trichloroaluminium,chloride aluminium
7446-70-0
AlCl3
133.34
aluminium chloride anhydrous,aluminum iii chloride,aluminiumchloride,aluminumchloride,aluminum chlorid,aluminum cloride,alumimum chloride,aluminum-chloride,trichloroaluminium,chloride aluminium
for ultra trace analysis, Lewis acid
Assay Percent Range 99.999%
Linear Formula AlCl3
Fieser 01,24; 02,21; 03,7; 04,10; 05,10; 06,17; 07,7; 08,13; 09,11; 10,9; 11,25; 12,26; 13,15; 14,20; 15,10; 16,10; 17,15
Merck Index 15,333
Solubility Information Solubility in water: reacts. Other solubilities: freely miscible with benzene, nitrobenzene,, benzophenone, carbon tetrachloride and chloroform
Formula Weight 133.34
Physical Form Powder
Percent Purity 99.999%
Grade Trace Metal Basis
Density 2.4400g/mL
Iron (Fe) 5ppm max.
Packaging Glass bottle
Solubility Solubility in water: reacts. Other solubilities: freely miscible with benzene, nitrobenzene,, benzophenone, carbon tetrachloride and chloroform
Specific Gravity 2.44
Total Trace Metal Impurities 0.001% max.
Color White
Melting Point 194.0°C
Quantity 100g
Chemical Name or Material Aluminium chloride
Show More Show Less

GHS H StatementCauses severe skin burns and eye damage.Reacts violently with water.

GHS P StatementWear protective gloves/protective clothing/eye protection/face protection.IF IN EYES: Rinse cautiously with water for several minutes.Remove contact lenses,if present and easy to do.Continue rinsing.Immediately call a POISON CENTER or doctor/physician.IF SWALLOWED: Rinse mouth.Do NOT induce vomiting.IF ON SKIN (or hair): Take off immediately all contaminated clothing.Rinse skin with water/shower.

;GHS P StatementWear protective gloves/protective clothing/eye protection/face protection.IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses,if present and easy to do. Continue rinsing.Immediately call a POISO