Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Aluminum, Fine Powder, Stabilized About 2% fat, MilliporeSigma™
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Metal can
Specifications
Appearance | Metallic powder |
Quantity | 250 g |
UN Number | UN 1396 |
Formula Weight | 26.98 |
Odor | Odorless |
Packaging | Metal can |
Form | Solid |
Chemical Name or Material | Aluminum |
Safety and Handling
Hazard Category | H228: Flammable solid. H261: In contact with water releases flammable gases. |
DOTInformation | DOT Class: 4.3, Packaging Group: II |
EINECSNumber | 231-072-3 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction