Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminium nitrate nonahydrate, ACS reagent, Acros Organics

Click to view available options
Quantity:
1kg
2.5kg
7784-27-2
AlN3O9·9H2O
375.13
SWCIQHXIXUMHKA-UHFFFAOYSA-N
aluminum nitrate nonahydrate,aluminum trinitrate nonahydrate,unii-8mc6621v1h,aluminum iii nitrate, nonahydrate 1:3:9,nitric acid aluminum salt,nitric acid, aluminum salt, nonahydrate 8ci,9ci,aluminum nitrate, acs,aluminium 3+ nonahydrate trinitronate,aluminum nitrate nonahydrate, low mercury, puratronic
24567
aluminum;trinitrate;nonahydrate
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Al+3]
7784-27-2
AlN3O9·9H2O
375.13
SWCIQHXIXUMHKA-UHFFFAOYSA-N
aluminum nitrate nonahydrate,aluminum trinitrate nonahydrate,unii-8mc6621v1h,aluminum iii nitrate, nonahydrate 1:3:9,nitric acid aluminum salt,nitric acid, aluminum salt, nonahydrate 8ci,9ci,aluminum nitrate, acs,aluminium 3+ nonahydrate trinitronate,aluminum nitrate nonahydrate, low mercury, puratronic
24567
aluminum;trinitrate;nonahydrate
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Al+3]
Linear Formula Al(NO3)3·9H2O
Merck Index 15,347
Solubility Information Solubility in water: 64 g/100mL (25°C). Other solubilities: soluble in alcohol, slightly soluble in acetone, practically insoluble in ethyl acetate and, pyridine
Formula Weight 375.13
Percent Purity 98.0 to 102.0% (Complexometry)
Grade ACS Reagent
Calcium (Ca) 0.005% max.
Chloride 0.001% max
Heavy Metals (as Pb) 0.001% max.
Insoluble Matter 0.005% max
Iron (Fe) 0.002% max.
Magnesium (Mg) 0.001% max.
Potassium (K) 0.002% max.
Sodium (Na) 0.005% max.
Solubility Solubility in water: 64 g/100mL (25°C). Other solubilities: soluble in alcohol, slightly soluble in acetone, practically insoluble in ethyl acetate and, pyridine
Sulfate 0.005% max
Melting Point 73.0°C
Quantity 1kg
Chemical Name or Material Aluminium nitrate nonahydrate
Show More Show Less

GHS H Statement
Causes serious eye damage.

GHS P Statement
Immediately call a POISON CENTRE or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger