Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminium oxide, for chromatography, basic, Brockmann I, 50-200 µm, 60A, ACROS Organics™

Click to view available options
Quantity:
1kg
5kg
25kg
Packaging:
Fibre drum
Plastic Bottle
1344-28-1
Al2O3
101.96
MFCD00003424
PNEYBMLMFCGWSK-UHFFFAOYSA-N
aluminium oxide,aluminum oxide,alpha-alumina,fasertonerde,abramant,abramax,abrarex,abrasit,aloxite,alundum
9989226
dialuminum;oxygen(2-)
[O-2].[O-2].[O-2].[Al+3].[Al+3]
1344-28-1
Al2O3
101.96
MFCD00003424
PNEYBMLMFCGWSK-UHFFFAOYSA-N
aluminium oxide,aluminum oxide,alpha-alumina,fasertonerde,abramant,abramax,abrarex,abrasit,aloxite,alundum
9989226
dialuminum;oxygen(2-)
[O-2].[O-2].[O-2].[Al+3].[Al+3]

Aluminium oxide, Al2O3, CAS Number-1344-28-1, abramant, abramax, abrarex, abrasit, aloxite, alpha-alumina, aluminium oxide, aluminum oxide, alundum, fasertonerde, 1kg, Na2O <0.4%, 2977.0 deg.C, White, 3.9700g/mL, 215-691-6, 300ppm max.

Aluminium oxide, Al2O3, CAS Number-1344-28-1, abramant, abramax, abrarex, abrasit, aloxite, alpha-alumina, aluminium oxide, aluminum oxide, alundum, fasertonerde, 1kg, Na2O <0.4%, 2977.0 deg.C, White, 3.9700g/mL, 215-691-6, 300ppm max.

For column chromatography
Additional Information Na2O <0.4%
Linear Formula Al2O3
Fieser 01,19; 02,17; 03,6; 04,8; 06,16; 07,5; 08,9; 09,8; 10,8; 11,22; 13,14; 15,9; 16,9
Merck Index 15, 350
Solubility Information Solubility in water: insoluble. Other solubilities: slightly sol. in alkaline sol.(forming hydroxides),practically insoluble in non-polar organic solvent,difficultly soluble in mineral acids
Formula Weight 101.96
Physical Form Powder
Grade For chromatography
Density 3.9700g/mL
Fe2O3 300ppm max.
Packaging Fibre drum
Silicon Dioxide 300ppm max.
Solubility Solubility in water: insoluble. Other solubilities: slightly sol. in alkaline sol.(forming hydroxides), practically insoluble in non-polar organic solvent, difficultly soluble in mineral acids
Specific Gravity 3.97
Surface Area 135 to 165 m2/g
Boiling Point 2977.0°C
Color White
Melting Point 2030.0°C
pH 9 to 10 (10% aq. suspension)
Quantity 25kg
Chemical Name or Material Aluminium oxide
Show More Show Less

EINECSNumber : 215-691-6