Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminium oxide, for chromatography, neutral, Brockmann I, 50-200 µm, 60A, Acros Organics

Click to view available options
Quantity:
1kg
2.5kg
25kg
1344-28-1
Al2O3
101.96
MFCD00003424
PNEYBMLMFCGWSK-UHFFFAOYSA-N
aluminium oxide,aluminum oxide,alpha-alumina,fasertonerde,abramant,abramax,abrarex,abrasit,aloxite,alundum
9989226
dialuminum;oxygen(2-)
[O-2].[O-2].[O-2].[Al+3].[Al+3]
1344-28-1
Al2O3
101.96
MFCD00003424
PNEYBMLMFCGWSK-UHFFFAOYSA-N
aluminium oxide,aluminum oxide,alpha-alumina,fasertonerde,abramant,abramax,abrarex,abrasit,aloxite,alundum
9989226
dialuminum;oxygen(2-)
[O-2].[O-2].[O-2].[Al+3].[Al+3]
for column chromatography
Linear Formula Al2O3
Fieser 01,19; 02,17; 03,6; 04,8; 06,16; 07,5; 08,9; 09,8; 10,8; 11,22; 13,14; 15,9; 16,9
Merck Index 15, 350
Solubility Information Solubility in water: insoluble
Formula Weight 101.96
Physical Form Fine Powder
Grade For chromatography
Density 3.97g/mL
Fe2O3 300ppm max.
Packaging Plastic bottle
Silicon Dioxide 300ppm max.
Sodium Oxide 0.4% max.
Solubility Solubility in water: insoluble.
Specific Surface Area 135 to 165 m2/g (BET)
Boiling Point 2980.0°C
Color White
Melting Point 2050.0°C
pH 6.5 to 7.5 (5% aq. suspension, stirred)
Quantity 2.5kg
Chemical Name or Material Aluminium oxide, Activated, Neutral, Brockmann I
Show More Show Less

EINECSNumber : 215-691-6