Alfa Aesar™ Aluminum 2,4-pentanedionate, 99.995+% (metals basis)

Manufacturer: Alfa Aesar™ 4453806

Quantity 5g

 View more versions of this product

Catalog No. AA4453806

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13963-57-0
    Molecular Formula C15H21AlO6
    Formula Weight 324.31
    MDL Number MFCD00000013
    UN Number UN3467
    Beilstein 4157942
    Synonym Aluminum acetylacetonate
    Chemical Name or Material Aluminum 2,4-pentanedionate

    Specifications

    Formula Weight 324.31
    Health Hazard 3 GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    Boiling Point 314° to 316°C
    Quantity 5g
    Beilstein 4157942
    RTECS Number BD2230000
    Molecular Formula C15H21AlO6
    MDL Number MFCD00000013
    CAS 13963-57-0
    UN Number UN3467
    Synonym Aluminum acetylacetonate
    Density 1.27g/mL
    Melting Point 192° to 194°C
    Physical Form Powder
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301-H315-H319-H335
    Toxic if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOMETALLIC COMPOUND, SOLID, TOXIC, N.O.S.
    Chemical Name or Material Aluminum 2,4-pentanedionate
    Assay ≥99.995%, (metals basis)

    GHS H Statement
    H301-H315-H319-H335
    Toxic if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit