Alfa Aesar™ Aluminum 2,4-pentanedionate

Manufacturer: Alfa Aesar™ 1254222

Quantity 100g

 View more versions of this product

Catalog No. AA1254222

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13963-57-0
    Formula Weight 324.31
    MDL Number MFCD00000013
    UN Number UN3467
    Beilstein 4157942
    Synonym Aluminum acetylacetonate; Tris(acetylacetonato)aluminum(III)
    Chemical Name or Material Aluminum 2,4-pentanedionate

    Specifications

    Formula Weight 324.31
    Health Hazard 3 GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Physical Form Powder
    UN Number UN3467
    Health Hazard 2 GHS H Statement
    H300-H315-H319-H335
    Fatal if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    RTECS Number BD2230000
    TSCA TSCA
    Health Hazard 1 Danger
    Density 1.27g/mL
    Melting Point 192° to 194°C
    DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ORGANOMETALLIC COMPOUND, SOLID, TOXIC, N.O.S.
    Boiling Point 314° to 316°C
    CAS 13963-57-0
    MDL Number MFCD00000013
    Quantity 100g
    Beilstein 4157942
    Synonym Aluminum acetylacetonate; Tris(acetylacetonato)aluminum(III)
    Chemical Name or Material Aluminum 2,4-pentanedionate

    GHS H Statement
    H300-H315-H319-H335
    Fatal if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit