Alfa Aesar™ Aluminum acetate, basic hydrate

Manufacturer: Alfa Aesar™ A1162036

Quantity 500g

 View more versions of this product

Catalog No. AAA1162036

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 142-03-0
    Molecular Formula approx (CH3CO2)2AlOH·xH2O
    MDL Number MFCD00008688
    Merck Index 14,344
    Chemical Name or Material Aluminum acetate, Basic hydrate

    Specifications

    TSCA TSCA
    CAS 142-03-0
    Merck Index 14,344
    Quantity 500g
    Molecular Formula approx (CH3CO2)2AlOH·xH2O
    MDL Number MFCD00008688
    Chemical Name or Material Aluminum acetate, Basic hydrate

    Description

    Applications
    Aluminum acetate, basic hydrate is used in medicine as an astringent and an antiseptic. It is also used as a mordant in dyeing, waterproofing and fireproofing agent. Further, it acts as a disinfectant in embalming and a dusting powder. In addition to this, it is used in antiperspirants.

    Solubility
    Insoluble in water

    Notes
    Incompatible with strong oxidizing agents.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit