Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminum Acetate, Dibasic, Stabilized with Boric Acid, Powder, Approx. 30%, Spectrum™

Click to view available options
Quantity:
100 lbs.
125 g
150 lbs.
2.5 kg
500 g
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
7360-44-3,10043-35-3
7360-44-3,10043-35-3

AL223, C2H5O4Al•H3BO3

Spectrum™ Aluminum Acetate, Dibasic, Stabilized with Boric Acid, Powder is a salt that is used as a topical astringent to treat skin irritations from poison ivy, substances such as soaps and cosmetics and insect bites. Aluminum acetate is particularly helpful for foot problems including athlete’s foot, swelling, and excessive sweating. It is also used to treat ear canal infections. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.
CAS Max % 0.3,0.7
Percent Purity Approx.30%
Chloride 0.0001
Heavy Metals (as Pb) 5e-005
Packaging Poly Pail
Quantity 2.5 kg