Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminum Ammonium Sulfate Dodecahydrate Products for Science Education

Click to view available options
Quantity:
100g
500g
Packaging:
Reagent Grade
7784-26-1
AlH28NO20S2
453.313
WZUKKIPWIPZMAS-UHFFFAOYSA-K
ammonia alum,unii-5c36drl9zn,aluminium ammonium sulfate dodecahydrate,aluminum ammonium disulfate dodecahydrate,ammonium aluminum sulfate hydrate,alum, ammonium usp,aluminum ammonium sulfate dodecahydrate,ammonium aluminum sulfate dodecahydrate,5c36drl9zn,ammonium aluminum disulfate dodecahydrate
62668
aluminum;azanium;disulfate;dodecahydrate
[NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Al+3]
7784-26-1
AlH28NO20S2
453.313
WZUKKIPWIPZMAS-UHFFFAOYSA-K
ammonia alum,unii-5c36drl9zn,aluminium ammonium sulfate dodecahydrate,aluminum ammonium disulfate dodecahydrate,ammonium aluminum sulfate hydrate,alum, ammonium usp,aluminum ammonium sulfate dodecahydrate,ammonium aluminum sulfate dodecahydrate,5c36drl9zn,ammonium aluminum disulfate dodecahydrate
62668
aluminum;azanium;disulfate;dodecahydrate
[NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Al+3]
Linear Formula AlNH4(SO4)2·12H2O
Formula Weight 453.32
Physical Form Solid
Grade Reagent
Recommended Applications Education
Density 1.6g/mL
Packaging Reagent Grade
Anhydrous or Hydrate Dodecahydrate
Color White
Quantity 100g
Chemical Name or Material Aluminum Ammonium Sulfate, Dodecahydrate
Show More Show Less

ShelfLife : 5 years