Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminum Ammonium Sulfate, Dodecahydrate, Powder, FCC, 99.5-100.5%, Spectrum™

Click to view available options
Quantity:
100 lbs.
12 kg
2.5 kg
500 g
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
7784-26-1
AlH28NO20S2
453.31
WZUKKIPWIPZMAS-UHFFFAOYSA-K
aluminium(3+) ammonium dodecahydrate disulfate
[NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[Al+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
7784-26-1
AlH28NO20S2
453.31
WZUKKIPWIPZMAS-UHFFFAOYSA-K
aluminium(3+) ammonium dodecahydrate disulfate
[NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[Al+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

AL224, 7784-26-1, AlNH4(SO4)2•12H2O

Spectrum™ Aluminum Ammonium Sulfate, Dodecahydrate, Powder, FCC is used in foods as a buffer and neutralizing agent. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 100%
Percent Purity 99.5 to 100.5%
Fluoride 0.003%
Lead (Pb) 3 mg/kg
Packaging Poly Pail
Selenium (Se) 0.003%
Quantity 2.5 kg