Alfa Aesar™ Aluminum arsenide, 99.5% (metals basis)

In rectifiers, transistors, thermistors

Manufacturer: Alfa Aesar™ 1452214

Quantity 25g

 View more versions of this product

Catalog No. AA1452214

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 22831-42-1
    Formula Weight 101.9
    MDL Number MFCD00046166
    UN Number UN1557
    Chemical Name or Material Aluminum arsenide

    Specifications

    Formula Weight 101.9
    Health Hazard 3 GHS P Statement
    P261-P301+P310-P304+P340-P311-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Call a POISON CENTER or doctor/physician.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Mesh Size −20 Mesh
    Physical Form Powder
    MDL Number MFCD00046166
    Quantity 25g
    Health Hazard 1 Danger
    Density 3.81g/mL
    DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ARSENIC COMPOUNDS, SOLID, N.O.S.
    TSCA TSCA
    CAS 22831-42-1
    UN Number UN1557
    Health Hazard 2 GHS H Statement
    H301-H331
    Toxic if swallowed.
    Toxic if inhaled.
    Melting Point 1740°C
    Chemical Name or Material Aluminum arsenide
    Assay 99.5%, (metals basis)

    GHS H Statement
    H301-H331
    Toxic if swallowed.
    Toxic if inhaled.

    GHS P Statement
    P261-P301+P310-P304+P340-P311-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Call a POISON CENTER or doctor/physician.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit