Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminium bromide, 98+%, extra pure, anhydrous, Acros Organics

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Glass bottle
7727-15-3
AlBr3
266.7
7727-15-3
AlBr3
266.7
Lewis acid
Assay Percent Range 98% min. (Complexometry)
Linear Formula AlBr3
Fieser 01,22; 02,19; 03,7; 04,10; 05,10; 06,17; 09,11
Merck Index 15,328
Solubility Information Solubility in water: reacts. Other solubilities: soluble in alcohol,carbon disulfide,benzene,,toluene,ether,nitrobenzene and xylene
Formula Weight 266.7
Physical Form Crystalline Solid
Percent Purity 98+%
Grade Extra Pure
Chloride 2000ppm max
Chlorine (Cl) 2000ppm max.
Iron (Fe) 50ppm max.
Lead (Pb) 50ppm max.
Packaging Glass bottle
Solubility Solubility in water: reacts. Other solubilities: soluble in alcohol, carbon disulfide, benzene,, toluene, ether, nitrobenzene and xylene
Sulfate 200ppm max.
Boiling Point 265.0°C
Color Gray or White
Melting Point 97.0°C
Quantity 100g
Chemical Name or Material Aluminium bromide
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Reacts violently with water.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

EINECSNumber : 231-779-7