Alfa Aesar™ Aluminum bromide, ultra dry, 99.999% (metals basis)

Manufacturer: Alfa Aesar™ 4371922

Quantity 100g

 View more versions of this product

Catalog No. AA4371922

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 7727-15-3
    Molecular Formula AlBr3
    Formula Weight 266.71
    MDL Number MFCD00003421
    UN Number UN1725
    Merck Index 14,332
    Chemical Name or Material Aluminum bromide, Ultra dry

    Specifications

    Physical Form Lumps
    Formula Weight 266.71
    Packaging Ampule
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Boiling Point 265°C
    Color White to orange
    Health Hazard 2 GHS H Statement
    H314-H318-H302
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Harmful if swallowed.
    Storage Note 1
    • Ampouled under argon
    Molecular Formula AlBr3
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: ALUMINUM BROMIDE, ANHYDROUS
    TSCA TSCA
    Particle Size 5 to 10mm
    MDL Number MFCD00003421
    Merck Index 14,332
    Melting Point 97.5°C
    Density 3.21g/mL
    Odor Pungent
    Sensitivity Moisture Sensitive
    CAS 7727-15-3
    UN Number UN1725
    RTECS Number BD0350000
    Quantity 100g
    Chemical Name or Material Aluminum bromide, Ultra dry
    Assay 99.999%, (metals basis)

    GHS H Statement
    H314-H318-H302
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Harmful if swallowed.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    • Ampouled under argon

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit