Learn More
Brady™ Aluminum Caution/ Precaucion Sign: CORROSIVE MATERIALS WEAR REQUIRED PROTECTION/ MATERIAL CORROSIVO USAR LA PROTECCION REQUERIDA
Improve workplace safety with proper signage. The Brady™ Caution/precaucion Sign is a durable, clear and effective tool to properly mark appropriate areas and to help prevent workplace injury.
$15.25 - $25.30
Specifications
Adhesive Type Non-adhesive
Color Black on Yellow
Message CORROSIVE MATERIALS WEAR REQUIRED PROTECTION/MATERIAL CORROSIVO USAR LA PROTECCION REQUERIDA
Quantity 1 Ea.
Shape Rectangular
View More Specs
Products 2
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity & Availability  
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity & Availability  
19-105-126
View Documents
Brady™
125367
7 x 10 in.
Each for $15.25
Add to cart
19-105-129
View Documents
Brady™
125370
10 x 14 in.
Each for $25.30
Add to cart
Description
Description
  • Aluminum material is abrasion-resistant and chemical-resistant
  • Aluminum signs give you the strength and durability of metal, but they never rust and can tolerate extreme temperatures, years of chemical spills, wash downs and cleanings
  • Safety signs are important facility visuals that are vital to communicating warnings and other safety information
Specifications
Specifications
Non-adhesive
CORROSIVE MATERIALS WEAR REQUIRED PROTECTION/MATERIAL CORROSIVO USAR LA PROTECCION REQUERIDA
Rectangular
Aluminum
Black on Yellow
1 Ea.
0.035 in.