Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Brady™ Aluminum Caution Sign: HAZARDOUS MATERIALS MAY BE USED IN THIS AREA SEE MSDS
Improve workplace safety with proper signage and markings. This Brady™ corner hole mounted sign is a durable, clear and effective tool to raise awareness to chemical, biohazard and hazardous material warnings.
$24.70 - $43.60
Specifications
Color | Black on Yellow |
---|---|
Message | HAZARDOUS MATERIALS MAY BE USED IN THIS AREA SEE MSDS |
Shape | Rectangular |
Temperature Range (English) | -40°F to 230°F |
Finish | Not Laminated |
Catalog Number | Mfr. No. | Dimensions (L x W) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Dimensions (L x W) | Price | Quantity & Availability | |||||
19-105-558
|
Brady™
126000 |
10 x 7 in. |
Each for $24.70
|
|
19-105-561
|
Brady™
126003 |
14 x 10 in. |
Each for $43.60
|
|
Description
- Aluminum material is abrasion-resistant and chemical-resistant
- Aluminum signs provide the strength and durability of metal, but never rust and can tolerate extreme temperatures, years of chemical spills, wash downs and cleanings
- Safety signs are important facility visuals that are vital to communicating warnings and other safety information
- For indoor and outdoor use
- Printed on polymer plastic and overlaminated with a clear plastic coating to protect graphics against abrasion, sunlight, chemicals, and weather up to 8 years outdoors, and even longer indoors
Specifications
Black on Yellow | |
Rectangular | |
Not Laminated | |
1 Ea. | |
230°F | |
0.035 in. |
HAZARDOUS MATERIALS MAY BE USED IN THIS AREA SEE MSDS | |
-40°F to 230°F | |
Aluminum | |
Corner holes | |
-40°F | |
Non-adhesive |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title