Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminum Chloride, 6.0 N, For Sulfide, APHA, Solution, Spectrum™

Click to view available options
Quantity:
125 mL
500 mL
Packaging:
Poly Bottle
7784-13-6,7732-18-5
7784-13-6,7732-18-5
Grade Chemical Solution
Packaging Poly Bottle
Quantity 500 mL

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.