Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminum Chloride, 99%, Reagent Grade, Honeywell™

Click to view available options
Quantity:
5g
100g
500g
Packaging:
Glass Bottle
7446-70-0
MFCD00003422
aluminium chloride anhydrous,aluminum iii chloride,aluminiumchloride,aluminumchloride,aluminum chlorid,aluminum cloride,alumimum chloride,aluminum-chloride,trichloroaluminium,chloride aluminium
7446-70-0
MFCD00003422
aluminium chloride anhydrous,aluminum iii chloride,aluminiumchloride,aluminumchloride,aluminum chlorid,aluminum cloride,alumimum chloride,aluminum-chloride,trichloroaluminium,chloride aluminium
Linear Formula AlCl3
UN Number UN1726
Formula Weight 133.34g/mol
Percent Purity 99%
Grade ReagentPlus™
Density 2.44g/cm3
Packaging Glass Bottle
Vapor Pressure 1mmHg (100°C)
Physical State Solid
Color White to Yellow
Melting Point 190°C
Quantity 500g
Chemical Name or Material Aluminum Chloride
Show More Show Less

P280-P305 + P351 + P338-P310

H314-H372-H373

DOTInformation : 8/PGII

EINECSNumber : 231-208-1

RTECSNumber : BD0525000