Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminum Chloride, Hexahydrate, Crystal, Spectrum™

Click to view available options
Quantity:
25 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
7784-13-6
7784-13-6
Grade Reagent
CAS Max % 1
Arsenic (As) 5e-06
Heavy Metals 5e-06
Insoluble Matter 0.0001
Iron (Fe) 5e-05
Packaging Poly Pail
Sulfate 0.0002
Quantity 12 kg

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.