Aluminum Chloride Hexahydrate (Crystalline/Certified), Fisher Chemical

Packaging Glass Bottle
Quantity 2.5kg

 View more versions of this product

Catalog No. A573-212

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 

Custom packing/Bulk ordering

You can now order this item in custom quantities.  Learn More

    Chemical Identifiers

    CAS 7784-13-6
    Physical Form Solid
    Molecular Formula AlCl3 · 6 H2O
    Formula Weight 241.43
    MDL Number MFCD00149134
    Chemical Name or Material Aluminum Chloride Hexahydrate
    Grade Certified Grade

    Specifications

    Formula Weight 241.43
    Melting Point 181°C
    Quantity 2.5kg
    DOT Information DOT Class 8, : Corrosive
    Physical Form Solid
    Color White
    pH 2.0
    Density 2.39g/mL
    Molecular Formula AlCl3 · 6 H2O
    Heavy Metals (as Pb) 5ppm max.
    Identification Pass Test
    Insoluble Matter 0.005% max.
    Iron (Fe) 5ppm max.
    Sulfate 0.010% max.
    MDL Number MFCD00149134
    ChemAlert Storage Symbol White
    Free Acid 0.10% max.
    Alkalis and Alkaline Earths 0.20% max.
    Packaging Glass Bottle
    Ammonia 0.010% max.
    CAS 7784-13-6
    Chemical Name or Material Aluminum Chloride Hexahydrate
    Grade Certified Grade
     

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling
  • Product Certifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit