Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminum Chloride Hexahydrate (Crystalline/USP), Fisher Chemical™

Catalog Number p-30749
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
7784-13-6
AlCl3H12O6
241.42
MFCD00149134
JGDITNMASUZKPW-UHFFFAOYSA-K
aluminium(3+) hexahydrate trichloride
O.O.O.O.O.O.[Al+3].[Cl-].[Cl-].[Cl-]
7784-13-6
AlCl3H12O6
241.42
MFCD00149134
JGDITNMASUZKPW-UHFFFAOYSA-K
aluminium(3+) hexahydrate trichloride
O.O.O.O.O.O.[Al+3].[Cl-].[Cl-].[Cl-]
Melting Point 181°C
Color White
pH 2.0
Physical Form Solid
Quantity 500 g
Formula Weight 241.43
Percent Purity 95.0 to 102.0% (USP)
Grade USP
Identification Pass Test (USP)
Packaging Poly Bottle
Density 2.39g/cm³
Chemical Name or Material Aluminum Chloride Hexahydrate
Show More Show Less
DOTInformation DOT Class 8, : Corrosive
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit