Aluminum Chloride Hexahydrate (Crystalline/USP), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.

Packaging Poly Bottle
Quantity 2.5kg

 View more versions of this product

Catalog No. A576-212

  Check Availability
Add to Cart

Custom packing/Bulk ordering

You can now order this item in custom quantities.  Learn More

    Chemical Identifiers

    CAS 7784-13-6
    Physical Form Solid
    Molecular Formula AlCl3 · 6 H2O
    Formula Weight 241.43
    MDL Number MFCD00149134
    Chemical Name or Material Aluminum Chloride Hexahydrate
    Assay 95.0 to 102.0% (USP)
    Grade USP

    Specifications

    Formula Weight 241.43
    Melting Point 181°C
    Quantity 2.5kg
    DOT Information DOT Class 8, : Corrosive
    Physical Form Solid
    Color White
    pH 2.0
    Density 2.39g/mL
    Molecular Formula AlCl3 · 6 H2O
    Heavy Metals (as Pb) 0.002% max. (USP)
    Identification Pass Test (USP)
    Iron (Fe) 0.001% max. (USP)
    Packaging Poly Bottle
    Residual Solvents Meets Requirements (USP)
    Sulfate Pass Test (USP)
    ChemAlert Storage Symbol White
    Alkalis and Alkaline Earths 0.5% max. (USP)
    MDL Number MFCD00149134
    CAS 7784-13-6
    Water 42.0 to 48.0% (USP)
    Chemical Name or Material Aluminum Chloride Hexahydrate
    Assay 95.0 to 102.0% (USP)
    Grade USP
     

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling
  • Product Certifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit