Alfa Aesar™ Aluminum chloride hexahydrate, Puratronic™, 99.9995% (metals basis)

Manufacturer: Alfa Aesar™ 10622A1

Quantity 1kg

 View more versions of this product

Catalog No. AA10622A1

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7784-13-6
    Formula Weight 241.43 (133.34 Anhydrous)
    MDL Number MFCD00149134
    UN Number UN3260
    Merck Index 14,337
    Chemical Name or Material Aluminum chloride hexahydrate
    Grade Puratronic™

    Specifications

    Formula Weight 241.43 (133.34 Anhydrous)
    Health Hazard 3 GHS P Statement
    P280-P303+P361+P353-P305+P351+P338-P310
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Immediately call a POISON CENTER or doctor/physician.
    TSCA TSCA
    MDL Number MFCD00149134
    UN Number UN3260
    Merck Index 14,337
    Physical Form Crystalline Aggregates
    Color White
    Density 2.398g/mL
    Odor Odorless
    Quantity 1kg
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
    RTECS Number BD0530000
    CAS 7784-13-6
    Melting Point 100°C (dec.)
    pH 2.5
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Chemical Name or Material Aluminum chloride hexahydrate
    Assay 99.9995%, (metals basis)
    Grade Puratronic™

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P280-P303+P361+P353-P305+P351+P338-P310
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Immediately call a POISON CENTER or doctor/physician.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit