Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminum Chloride, Hexahydrate, USP, 95-102%, Spectrum™

Click to view available options
Quantity:
100 g
500 g
2.5 kg
12 kg
25 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
7784-13-6
AlCl3H12O6
241.42
JGDITNMASUZKPW-UHFFFAOYSA-K
aluminium(3+) hexahydrate trichloride
O.O.O.O.O.O.[Al+3].[Cl-].[Cl-].[Cl-]
7784-13-6
AlCl3H12O6
241.42
JGDITNMASUZKPW-UHFFFAOYSA-K
aluminium(3+) hexahydrate trichloride
O.O.O.O.O.O.[Al+3].[Cl-].[Cl-].[Cl-]

AL225, 7784-13-6, AlCl3•6H2O

Spectrum™ Aluminum Chloride, Hexahydrate, USP is mainly used in topical formulations to treat hyperhidrosis (excessive sweating). All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 1
Percent Purity 95 to 102%
Packaging Poly Bottle
Quantity 500 g