Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminum Chlorohydrate, Dihydrate, USP, Spectrum™

Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
12359-72-7
12359-72-7

A1081, 12359-72-7, Aly(OH)(3y-z)Clz•2H2O

Spectrum™ Aluminum Chlorohydrate, Dihydrate, USP is a group of specific aluminum salts used in topical skin products (deodorants and antiperspirants) as an astringent in the management of hyperhidrosis. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 1
Arsenic (As) 2 ppm
Packaging Fiber Drum
pH Range 3.0-5.0
Quantity 50 kg