Alfa Aesar™ Aluminum Cobalt powder, -100 mesh, 99% (metals basis)

Manufacturer: Alfa Aesar™ 8832022

Quantity 100g

 View more versions of this product

Catalog No. AA8832022

  Check Availability
Add to Cart

    Chemical Identifiers

    MDL Number MFCD00239520
    UN Number UN3132
    Chemical Name or Material Aluminum Cobalt powder

    Specifications

    Health Hazard 2 GHS H Statement
    H250-H261-H228-H334-H317
    Catches fire spontaneously if exposed to air.
    In contact with water releases flammable gas.
    Flammable solid.
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    May cause an allergic skin reaction.
    Health Hazard 3 GHS P Statement
    P210-P222-P231+P232-P342+P311-P422a-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Do not allow contact with air.
    Handle under inert gas.
    Protect from moisture.
    If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
    Store contents under inert gas.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Mesh Size −100 Mesh
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: WATER-REACTIVE SOLID, FLAMMABLE, N.O.S.
    Avg. Mol. Wt. or Mol. Wt. Range Al:Co; 69:31 wt%
    TSCA TSCA
    Physical Form Powder
    MDL Number MFCD00239520
    UN Number UN3132
    Odor Odorless
    Quantity 100g
    Chemical Name or Material Aluminum Cobalt powder
    Assay 99%, (metals basis)

    GHS H Statement
    H250-H261-H228-H334-H317
    Catches fire spontaneously if exposed to air.
    In contact with water releases flammable gas.
    Flammable solid.
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    May cause an allergic skin reaction.

    GHS P Statement
    P210-P222-P231+P232-P342+P311-P422a-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Do not allow contact with air.
    Handle under inert gas.
    Protect from moisture.
    If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
    Store contents under inert gas.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit