Learn More
Thermo Scientific™ Aluminum Crimp Seals with Prefitted Septa for 12 x 32mm Vials Available on GSA/VA Contract for Federal Government customers only.
You can count on a secure, leak-resistant seal with Thermo Scientific™ National Aluminum Crimp Seals.
Manufacturer:  Thermo Scientific™ C40115A
 View more versions of this product
Catalog No. 03-378-343
  Check Availability
Add to cart
Description
Description
You can count on a secure, leak-resistant seal with Thermo Scientific™ National Aluminum Crimp Seals.
  • Anodized aluminum seals fit all 11mm crimp-top vials
  • Seal 2000™ material is specially formulated for improved background performance over PTFE/red rubber
  • Viton™ septa perform well with ECD detectors and are resistant to many aggressive solvents
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Black Viton Elastomer Septa; Silver Aluminum Seal
Black (Septa)þSilver (Cap)
Black Viton Elastomer Septa
Crimp Cap with Septum
C4011-5A
Silver
Aluminum (Cap)þViton™ Elastomer (Septa)
11mm
12 x 32 Vials
National™