Learn More
Thermo Scientific™ Aluminum Crimp Seals with Prefitted Septa for 8mm Crimp Top Vials Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ National Aluminum Crimp Seals provide a secure, leak-resistant seal that you can count on.
Manufacturer:  Thermo Scientific™ C4008-98G
 View more versions of this product
Catalog No. 06-406-19X
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications
Description
Thermo Scientific™ National Aluminum Crimp Seals provide a secure, leak-resistant seal that you can count on.
  • Available in a variety of colors and septum types
  • Aluminum seals must be applied with a crimping tool
Description & Specifications
Specifications
8mm Crimp Top Vials
Clear PTFE/Red Rubber septa; Green
Green
8mm
General-purpose resealable septum
C4008-98G
Clear PTFE/Red Rubber septa