Learn More
Thermo Scientific Aluminum Crimp Seals with Prefitted Septa for 8mm Crimp Top Vials
Thermo Scientific™ National Aluminum Crimp Seals provide a secure, leak-resistant seal that you can count on.
Manufacturer:  Thermo Scientific C400898Y
 View more versions of this product
Catalog No. 03-377-416
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Description
Thermo Scientific™ National Aluminum Crimp Seals provide a secure, leak-resistant seal that you can count on.
  • Available in a variety of colors and septum types
  • Aluminum seals must be applied with a crimping tool
Description & Specifications
Specifications
8mm Crimp Top Vials
Clear PTFE/red rubber septa; Yellow
Yellow
8mm
General-purpose resealable septum
C4008-98Y
Clear PTFE/red rubber septa