Learn More
METTLER TOLEDO 3P INDUSTRIAL ALUMINUM CRUCIBLE 40 100 PK DFS Item
Manufacturer:  METTLER TOLEDO 3P INDUSTRIAL 26763
Aluminum Crucible standard, 40 ul, w/o pin, with lid The precisely machined crucibles with strong and flat crucible bases guarantee excellent thermal contact between the crucible and the sensor and low temperature gradients. Hermetically sealed crucibles withstand pressures of up to 0.2 MPa and temperatures up to 640 Deg.C. The crucibles can be used with different types of lids: lids with 50 um holes for measurements in a self-generated atmosphere - piercing lids prevent samples stored in the autosampler from losing volatile constituents prior to measurements. Using a piercing lid and lid piercing kit, the crucibles are automatically pierced before being transferred to the measuring cell.
Catalog No. NC0100384