Learn More
Brady™ Aluminum Danger Sign: COMBUSTIBLE LIQUIDS.
Improve workplace safety with proper signage and markings. This Brady™ corner hole mounted sign is a durable, clear and effective tool to raise awareness to chemical, biohazard and hazardous material warnings.
$24.70 - $43.60
Specifications
Shape | Rectangular |
---|---|
Displayed Text | COMBUSTIBLE LIQUIDS. |
Temperature Range (English) | -40°F to 230°F |
Finish | Not Laminated |
Material | Aluminum |
Catalog Number | Mfr. No. | Dimensions (L x W) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Dimensions (L x W) | Price | Quantity & Availability | |||||
19-106-423
|
Brady™
144917 |
14 x 10 in. |
Each for $43.60
|
|
|||||
19-106-422
|
Brady™
144916 |
10 x 7 in. |
Each for $24.70
|
|
|||||
Description
- Aluminum material is abrasion-resistant and chemical-resistant
- Aluminum signs provide the strength and durability of metal, but never rust and can tolerate extreme temperatures, years of chemical spills, wash downs and cleanings
- Bold and bright, providing high visibility to critical messages
- For indoor and outdoor use
- Printed on polymer plastic and overlaminated with a clear plastic coating to protect graphics against abrasion, sunlight, chemicals, and weather up to 8 years outdoors, and even longer indoors
Specifications
Rectangular | |
-40°F to 230°F | |
Aluminum | |
Non-adhesive | |
230°F | |
0.035 in. |
COMBUSTIBLE LIQUIDS. | |
Not Laminated | |
1 Ea. | |
Corner holes | |
-40°F |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.