Learn More
Brady™ Aluminum Danger Sign: POISON KEEP OUT
Improve workplace safety with proper signage. The Brady™ Danger Sign is a durable, clear and effective tool to properly mark appropriate areas and to help prevent workplace injury.
$18.86 - $33.38
Specifications
Adhesive Type Non-adhesive
Color Black/Red on White
Message POISON KEEP OUT
Quantity 1 Ea.
Shape Rectangular
View More Specs
Products 2
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity & Availability  
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity & Availability  
19-105-870
View Documents
Brady™
126375
14 x 10 in.
Each for $33.38
Add to cart
19-105-867
View Documents
Brady™
126372
10 x 7 in.
Each for $18.86
Add to cart
Description
Description
  • Aluminum material is abrasion-resistant and chemical-resistant
  • Aluminum signs give you the strength and durability of metal, but they never rust and can tolerate extreme temperatures, years of chemical spills, wash downs and cleanings
  • Safety signs are important facility visuals that are vital to communicating warnings and other safety information
Specifications
Specifications
Non-adhesive
POISON KEEP OUT
Rectangular
Aluminum
Black/Red on White
1 Ea.
0.035 in.