Learn More
Brady™ Aluminum Danger Sign: POISONOUS GAS STORAGE
Improve workplace safety with proper signage. The Brady™ Danger Sign is a durable, clear and effective tool to properly mark appropriate areas and to help prevent workplace injury.
Specifications
Adhesive Type Non-adhesive
Color Black/Red on White
Message POISONOUS GAS STORAGE
Quantity 1 Ea.
Shape Rectangular
View More Specs
Products 2
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity & Availability  
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity & Availability  
19-105-855
View Documents
Brady™
126360
10 x 7 in. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
19-105-858
View Documents
Brady™
126363
14 x 10 in. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
  • Aluminum material is abrasion-resistant and chemical-resistant
  • Aluminum signs give you the strength and durability of metal, but they never rust and can tolerate extreme temperatures, years of chemical spills, wash downs and cleanings
  • Safety signs are important facility visuals that are vital to communicating warnings and other safety information
Specifications
Specifications
Non-adhesive
POISONOUS GAS STORAGE
Rectangular
Aluminum
Black/Red on White
1 Ea.
0.035 in.