Learn More
Aluminum dihydrogen phosphate, 50% w/w aqueous solution, Thermo Scientific Chemicals
Description
Aluminum dihydrogen phosphate is used as intermediates and solid separation agents. It is also used in petroleum, chemical industry, shipbuilding, space technology, electric industry, high-temperature kilns, heat treatment resistance furnaces and electric insulation. Further, it is used as inorganic coating.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Colorless |
Physical Form | Liquid |
Quantity | 1 kg |
Assay Percent Range | 50% w/w aqueous solution |
Solubility Information | Soluble in water and ethanol. Insoluble in acetone. |
Formula Weight | 317.94 |
Appearance | Colorless liquid |
Density | 1.5 |
Chemical Name or Material | Aluminum dihydrogen phosphate |
Safety and Handling
EINECSNumber | 215-691-6 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.