Alfa Aesar™ Aluminum Dish, Flat Bottom and Straight Sides
Manufacturer: Alfa Aesar™ 39069UD
 View more versions of this product
Catalog No. AA39069UD
  Check Availability
Add to Cart
    Specifications
    Diameter (Metric) 51mm
    Height (Metric) 12.5mm
    Shape Flat bottom and straight sides
    Material Aluminum
  • Description & Specifications
Related Products
Product Content Feedback