Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminum etchant, Alfa Aesar™

Standard Aluminum Etchant For Use On Silicon Devices and Other Microelectronic Applications

Manufacturer:  Alfa Aesar 44581R0

Quantity 1 qt.

 View more versions of this product

Catalog No. AA44581R0

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

Liquid
MFCD08064399
Aluminum etchant

Specifications

1 qt.
Liquid
Aluminum etchant
1.45g/cm3 at 20°C
MFCD08064399

Safety and Handling

SDS & Safety Documents

H314-H318: Causes severe skin burns and eye damage. Causes serious eye damage.

DOTInformation : DOT Class: 8, Packaging Group: II

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit