Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Aluminum fluoride, anhydrous, 99+%

Manufacturer:  Alfa Aesar™ 1149718

Quantity 50g

 View more versions of this product

Catalog No. AA1149718


  Check Availability
Add to cart

Specifications

Description

  • In ceramics, as flux in metallurgy, in aluminum manufacture, as inhibitor of fermentation, as catalyst in organic reactions
Description & Specifications

Chemical Identifiers

7784-18-1
MFCD00003426
Aluminum fluoride, Anhydrous
83.98
14,339

Specifications

7784-18-1
83.98
14,339
2.882g/mL
1291°C (subl.)
Aluminum fluoride, Anhydrous
Powder
MFCD00003426
Practically insoluble in water; Sparingly soluble in acids and alkalies
Hygroscopic
50g
≥99%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H318-H302-H335-H315
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

RTECSNumber : BD0725000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit