Alfa Aesar™ Aluminum fluoride, anhydrous, 99.5%

For vacuum deposition

Manufacturer: Alfa Aesar™ 3633506

Quantity 5g

 View more versions of this product

Catalog No. AA3633506

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7784-18-1
    Formula Weight 83.98
    MDL Number MFCD00003426
    Merck Index 14,339
    Chemical Name or Material Aluminum fluoride, Anhydrous

    Specifications

    Formula Weight 83.98
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Particle Size 3 to 12mm
    Merck Index 14,339
    Quantity 5g
    Health Hazard 2 GHS H Statement
    H318-H302-H335-H315
    Causes serious eye damage.
    Harmful if swallowed.
    May cause respiratory irritation.
    Causes skin irritation.
    TSCA TSCA
    Physical Form Sintered Lump
    Sensitivity Hygroscopic
    MDL Number MFCD00003426
    CAS 7784-18-1
    Melting Point 1291°C (subl.)
    Health Hazard 1 Danger
    Density 2.882g/mL
    RTECS Number BD0725000
    Chemical Name or Material Aluminum fluoride, Anhydrous
    Assay 99.5%

    GHS H Statement
    H318-H302-H335-H315
    Causes serious eye damage.
    Harmful if swallowed.
    May cause respiratory irritation.
    Causes skin irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit