Alfa Aesar™ Aluminum fluoride hydrate, Puratronic™, 99.99% (metals basis)

Manufacturer: Alfa Aesar™ 1062314

Quantity 25g

 View more versions of this product

Catalog No. AA1062314

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 32287-65-3
    Formula Weight 83.98 (Anhydrous)
    MDL Number MFCD00003426
    UN Number UN3260
    Chemical Name or Material Aluminum fluoride hydrate
    Grade Puratronic™

    Specifications

    Formula Weight 83.98 (Anhydrous)
    Health Hazard 3 GHS P Statement
    P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Mesh Size −22 Mesh
    UN Number UN3260
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
    Physical Form Powder
    MDL Number MFCD00003426
    CAS 32287-65-3
    Quantity 25g
    TSCA TSCA
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H300-H314-H318
    Fatal if swallowed.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Chemical Name or Material Aluminum fluoride hydrate
    Assay 99.99%, (metals basis)
    Grade Puratronic™

    GHS H Statement
    H300-H314-H318
    Fatal if swallowed.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit